4-Bromo-2-fluoroanisole 98%

Supplier: Apollo Scientific
Warning

PC1398-10G
APOSPC1398-10GEA 21.9 EUR
APOSPC1398-10G
4-Bromo-2-fluoroanisole 98%
4-Bromo-2-fluoroanisole
Formula: C₇H₆BrFO
Boiling Pt: 208 °C
Density: 1,59 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00011710
CAS Number: 2357-52-0
EINECS: 219-096-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR