4-Fluoropyridine hydrochloride 95%

Supplier: Apollo Scientific
Warning

PC1406-1G PC1406-5G PC1406-250MG
APOSPC1406-1GEA 168 EUR
APOSPC1406-1G APOSPC1406-5G APOSPC1406-250MG
4-Fluoropyridine hydrochloride 95%
4-Fluoropyridine hydrochloride
Formula: C₅H₅ClFN
MW: 133.55 g/mol
Melting Pt: 110 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00234055
CAS Number: 39160-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR