1-Bromo-3-fluoro-4-iodobenzene 97%

Supplier: Apollo Scientific
Warning

PC1426F-5G PC1426F-25G PC1426F-100G PC1426F-500G
APOSPC1426F-5GEA 0 EUR
APOSPC1426F-5G APOSPC1426F-25G APOSPC1426F-100G APOSPC1426F-500G
1-Bromo-3-fluoro-4-iodobenzene 97%
1-Bromo-3-fluoro-4-iodobenzene
Formula: C₆H₃BrFI
Boiling Pt: 48…51 °C
Melting Pt: 48…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010608
CAS Number: 105931-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR