5-Fluoro-2-pyridineboronic acid 96%

Supplier: Apollo Scientific
Warning

PC1428-1G PC1428-250MG
APOSPC1428-1GEA 393 EUR
APOSPC1428-1G APOSPC1428-250MG
5-Fluoro-2-pyridineboronic acid 96%
5-Fluoro-2-pyridineboronic acid
Formula: C₅H₅BFNO₂
Storage Temperature: Freezer
MDL Number: MFCD08705712
CAS Number: 946002-10-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR