2,6-Difluorophenylhydrazine

Supplier: Apollo Scientific

Synonyms: (2,6-Difluorophenyl)hydrazine

PC1502-1G
APOSPC1502-1GEA 68.3 EUR
APOSPC1502-1G
2,6-Difluorophenylhydrazine
2,6-Difluorophenylhydrazine
Formula: C₆H₆F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD06658266
CAS Number: 119452-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR