3,4-Dichloro-6-(trifluoromethyl)toluene

Supplier: Apollo Scientific
Warning

Synonyms: 4,5-Dichloro-2-methylbenzotrifluoride

PC1535-1G PC1535-25G PC1535-5G PC1535-100G
APOSPC1535-1GEA 0 EUR
APOSPC1535-1G APOSPC1535-25G APOSPC1535-5G APOSPC1535-100G
3,4-Dichloro-6-(trifluoromethyl)toluene
3,4-Dichloro-6-(trifluoromethyl)toluene
Formula: C₈H₅Cl₂F₃
Boiling Pt: 97…100 °C/23mmHg
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD06246858
CAS Number: 74483-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR