3-Fluorothioanisole

Supplier: Apollo Scientific
Warning

PC1579-100G PC1579-25G PC1579-5G
APOSPC1579-100GEA 696 EUR
APOSPC1579-100G APOSPC1579-25G APOSPC1579-5G
3-Fluorothioanisole
3-Fluorothioanisole
Formula: C₇H₇FS
Storage Temperature: Ambient
MDL Number: MFCD03789131
CAS Number: 658-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR