4-Bromo-6-(trifluoromethyl)benzimidazole 97%

Supplier: Apollo Scientific
Warning

PC1594E-1G
APOSPC1594E-1GEA 43.5 EUR
APOSPC1594E-1G
4-Bromo-6-(trifluoromethyl)benzimidazole 97%
4-Bromo-6-(trifluoromethyl)benzimidazole
Formula: C₈H₄BrF₃N₂
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00067734
CAS Number: 175135-14-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR