2-Bromo-3-(trifluoromethyl)pyridine 97%

Supplier: Apollo Scientific
Danger

PC1598E-25G PC1598E-5G
APOSPC1598E-25GEA 94.4 EUR
APOSPC1598E-25G APOSPC1598E-5G
2-Bromo-3-(trifluoromethyl)pyridine 97%
2-Bromo-3-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
Boiling Pt: 76 °C/8mmHg
Melting Pt: 37…41 °C
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00153084
CAS Number: 175205-82-0
UN: 2926
ADR: 4.1+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR