3-Fluoro-5-methoxyphenylboronic acid 97%

Supplier: Apollo Scientific
Warning

PC1621-25G PC1621-5G PC1621-100G
APOSPC1621-25GEA 400 EUR
APOSPC1621-25G APOSPC1621-5G APOSPC1621-100G
3-Fluoro-5-methoxyphenylboronic acid 97%
3-Fluoro-5-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD07363780
CAS Number: 609807-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR