Potassium-4-tert-butylphenyltrifluoroborate 96%

Supplier: Apollo Scientific

PC1752-25G PC1752-5G
APOSPC1752-25GEA 181 EUR
APOSPC1752-25G APOSPC1752-5G
Potassium-4-tert-butylphenyltrifluoroborate 96%
Potassium 4-tert-butylphenyltrifluoroborate
Formula: C₁₀H₁₃BF₃K
Melting Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD06659940
CAS Number: 423118-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR