Potassium 6-chloropyridine-3-trifluoroborate

Supplier: Apollo Scientific

PC200191-1G PC200191-500MG PC200191-5G
APOSPC200191-1GEA 168 EUR
APOSPC200191-1G APOSPC200191-500MG APOSPC200191-5G
Potassium 6-chloropyridine-3-trifluoroborate
Potassium 6-chloropyridine-3-trifluoroborate
Formula: C₅H₃BClF₃N·K
Storage Temperature: Ambient
MDL Number: MFCD09993185
CAS Number: 1235099-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR