3-Bromo-4-fluoro-7-nitro-1H-indazole

Supplier: Apollo Scientific

PC200290-1G PC200290-500MG
APOSPC200290-1GEA 626 EUR
APOSPC200290-1G APOSPC200290-500MG
3-Bromo-4-fluoro-7-nitro-1H-indazole
3-Bromo-4-fluoro-7-nitro-1H-indazole
Formula: C₇H₃BrFN₃O₂
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD20444429
CAS Number: 1353877-95-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR