N-(3,5-Difluorophenyl)-2,2,2-trifluoroacetamide

Supplier: Apollo Scientific

PC200428-1G
APOSPC200428-1GEA 538 EUR
APOSPC200428-1G
N-(3,5-Difluorophenyl)-2,2,2-trifluoroacetamide
N-(3,5-Difluorophenyl)-2,2,2-trifluoroacetamide
Formula: C₈H₄F₅NO
Storage Temperature: Ambient
MDL Number: MFCD20546924
CAS Number: 1378573-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR