Methyl 4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

Supplier: Apollo Scientific

PC200431-1G
APOSPC200431-1GEA 201 EUR
APOSPC200431-1G
Methyl 4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Methyl 4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Formula: C₁₄H₁₀F₃NO₃
MW: 297.229 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD27957294
CAS Number: 869109-13-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR