Methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methoxy}benzoate

Supplier: Apollo Scientific

PC200432-1G
APOSPC200432-1GEA 529 EUR
APOSPC200432-1G
Methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methoxy}benzoate
Methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methoxy}benzoate
Formula: C₁₅H₁₁ClF₃NO₃
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD27957299
CAS Number: 1565827-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR