5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine

Supplier: Apollo Scientific

PC200519-1G
APOSPC200519-1GEA 239 EUR
APOSPC200519-1G
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
5-(4-Fluorobenzyl)-1,3,4-thiadiazol-2-amine
Formula: C₉H₈FN₃S
Storage Temperature: Ambient
MDL Number: MFCD01364163
CAS Number: 39181-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR