Methyl-4-fluoro-2-methoxybenzoate

Supplier: Apollo Scientific
Warning

PC200606-1G PC200606-5G PC200606-10G
APOSPC200606-1GEA 21.9 EUR
APOSPC200606-1G APOSPC200606-5G APOSPC200606-10G
Methyl-4-fluoro-2-methoxybenzoate
Methyl 4-fluoro-2-methoxybenzoate
Formula: C₉H₉FO₃
Boiling Pt: 78…80 °C/0,05mmHg
Storage Temperature: Ambient
MDL Number: MFCD06203990
CAS Number: 204707-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR