5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 96%

Supplier: Apollo Scientific

PC201110-1G
APOSPC201110-1GEA 156 EUR
APOSPC201110-1G
5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 96%
5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one
Formula: C₉H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD18208171
CAS Number: 127425-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR