6-Fluorobenzo[d]isoxazol-3-ylamine 95+%

Supplier: Apollo Scientific

PC201176-250MG
APOSPC201176-250MGEA 129 EUR
APOSPC201176-250MG
6-Fluorobenzo[d]isoxazol-3-ylamine 95+%
6-Fluorobenzo[d]isoxazol-3-ylamine
Formula: C₇H₅FN₂O
Storage Temperature: Ambient
MDL Number: MFCD09999138
CAS Number: 177995-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR