Ethyl 2,5-difluorobenzoate 95+%

Supplier: Apollo Scientific

PC201287-5G
APOSPC201287-5GEA 357 EUR
APOSPC201287-5G
Ethyl 2,5-difluorobenzoate 95+%
Ethyl 2,5-difluorobenzoate
Formula: C₉H₈F₂O₂
MDL Number: MFCD06204339
CAS Number: 708-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR