3-Fluoro-N-methylbenzylamine 96%

Supplier: Apollo Scientific
Danger

PC201369-25G
APOSPC201369-25GEA 148 EUR
APOSPC201369-25G
3-Fluoro-N-methylbenzylamine 96%
3-Fluoro-N-methylbenzylamine
Formula: C₈H₁₀FN
Boiling Pt: 183…184 °C
Density: 1,015 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04623547
CAS Number: 90389-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR