3-Bromo-5-(trifluoromethyl)picolinic acid 95%

Supplier: Apollo Scientific

PC201386-100MG PC201386-250MG PC201386-1G
APOSPC201386-100MGEA 111 EUR
APOSPC201386-100MG APOSPC201386-250MG APOSPC201386-1G
3-Bromo-5-(trifluoromethyl)picolinic acid 95%
3-Bromo-5-(trifluoromethyl)picolinic acid
Formula: C7H3BrF3NO2
Storage Temperature: Ambient
MDL Number: MFCD11036124
CAS Number: 959245-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR