Copper(II) trifluoromethanesulphonate 98%

Supplier: Apollo Scientific
Danger

PC2096E-100G PC2096E-25G PC2096E-5G
APOSPC2096E-100GEA 105 EUR
APOSPC2096E-100G APOSPC2096E-25G APOSPC2096E-5G
Copper(II) trifluoromethanesulphonate 98%
Copper (II) trifluoromethanesulphonate

Can produce alkenes at ambient temperatures by catalytic dehydration of alcohols and diols: Helv. Chim. Acta, 70, 607 (1987)

Formula: C₂CuF₆O₆S₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00077492
CAS Number: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR