(4-Chlorophenyl)trifluoroboranuide tetrabutylazanium

Supplier: Apollo Scientific

PC210020-1G PC210020-5G
APOSPC210020-1GEA 102 EUR
APOSPC210020-1G APOSPC210020-5G
(4-Chlorophenyl)trifluoroboranuide tetrabutylazanium
(4-Chlorophenyl)trifluoroboranuide tetrabutylazanium
Formula: C₆H₄BClF₃·N(C₄H₉)₄
MW: 421.819 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28101517
CAS Number: 411206-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR