4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC2389-1G PC2389-5G PC2389-10G
APOSPC2389-1GEA 21.9 EUR
APOSPC2389-1G APOSPC2389-5G APOSPC2389-10G
4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%
4-Methoxy-3-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD01091012
CAS Number: 50823-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR