1,2-Dichlorohexafluorocyclopentene 97%

Supplier: Apollo Scientific
Danger

PC2470-25G PC2470-5G
APOSPC2470-25GEA 427 EUR
APOSPC2470-25G APOSPC2470-5G
1,2-Dichlorohexafluorocyclopentene 97%
1,2-Dichlorohexafluorocyclopentene
Formula: C₅Cl₂F₆
Boiling Pt: 90 °C
Melting Pt: –105,8 °C
Density: 1,635 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019302
CAS Number: 706-79-6
EINECS: 211-895-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR