Pentafluoroanilinium trifluoromethanesulphonate 95%

Supplier: Apollo Scientific
Danger

PC2657-1G
APOSPC2657-1GEA 85.7 EUR
APOSPC2657-1G
Pentafluoroanilinium trifluoromethanesulphonate 95%
Pentafluoroanilinium trifluoromethanesulphonate
Formula: C₇H₃F₈NO₃S
Melting Pt: 300…302 °C
Storage Temperature: Ambient
MDL Number: MFCD08276416
CAS Number: 912823-79-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR