3-fluoro-5-iodopyridine

Supplier: Apollo Scientific
Warning

PC2676-1G PC2676-250MG
APOSPC2676-1GEA 29 EUR
APOSPC2676-1G APOSPC2676-250MG
3-fluoro-5-iodopyridine
3-Fluoro-5-iodopyridine
Formula: C₅H₃FIN
Storage Temperature: Refrigerator
MDL Number: MFCD08457780
CAS Number: 757950-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR