4-Bromo-3-(trifluoromethyl)benzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC27926-1G PC27926-250MG
APOSPC27926-1GEA 609 EUR
APOSPC27926-1G APOSPC27926-250MG
4-Bromo-3-(trifluoromethyl)benzyl fluoride 95%
4-Bromo-3-(trifluoromethyl)benzyl fluoride
Formula: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31664906
CAS Number: 2244084-02-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR