2,2-Difluoroethyl acetate 97%

Supplier: Apollo Scientific
Danger

PC2808-25G PC2808-5G PC2808-100G PC2808-1G
APOSPC2808-25GEA 364 EUR
APOSPC2808-25G APOSPC2808-5G APOSPC2808-100G APOSPC2808-1G
2,2-Difluoroethyl acetate 97%
2,2-Difluoroethyl acetate
Formula: C₄H₆F₂O₂
Boiling Pt: 105 °C
Density: 1,2025 g/cm³ (25 °C)
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00069133
CAS Number: 1550-44-3
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR