2,4-Difluoro-6-nitroaniline 98%

Supplier: Apollo Scientific
Warning

PC2849T-100G PC2849T-25G PC2849T-5G
APOSPC2849T-100GEA 184 EUR
APOSPC2849T-100G APOSPC2849T-25G APOSPC2849T-5G
2,4-Difluoro-6-nitroaniline 98%
2,4-Difluoro-6-nitroaniline
Formula: C₆H₄F₂N₂O₂
Melting Pt: 85…87 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00034058
CAS Number: 364-30-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR