Perfluorodecane 95%

Supplier: Apollo Scientific
Warning

Synonyms: Docosafluorodecane

PC2903-25G PC2903-5G PC2903-1G
APOSPC2903-25GEA 210 EUR
APOSPC2903-25G APOSPC2903-5G APOSPC2903-1G
Perfluorodecane 95%
Perfluorodecane
Formula: C₁₀F₂₂
Boiling Pt: 144,2 °C
Melting Pt: 36 °C
Density: 1,77 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01736060
CAS Number: 307-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR