1,1,1-Trifluorobutane 97%

Supplier: Apollo Scientific
Warning

PC2909-5G
APOSPC2909-5GEA 690 EUR
APOSPC2909-5G
1,1,1-Trifluorobutane 97%
1,1,1-Trifluorobutane
Formula: C₄H₇F₃
Boiling Pt: 16,74 °C
Melting Pt: –114,79 °C
Density: 1,014 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 460-34-4
UN: 3161
ADR: 2.1,

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR