4-Bromomethyl-2'-fluorobiphenyl

Supplier: Apollo Scientific

PC300621-1G
APOSPC300621-1GEA 433 EUR
APOSPC300621-1G
4-Bromomethyl-2'-fluorobiphenyl
4-Bromomethyl-2'-fluorobiphenyl
Formula: C₁₃H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD15146811
CAS Number: 193013-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR