N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea

Supplier: Apollo Scientific

PC300821-1G PC300821-5G
APOSPC300821-1GEA 125 EUR
APOSPC300821-1G APOSPC300821-5G
N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea
N-(4-Chlorophenyl)-N'-(4-fluorophenyl)urea
Formula: C₁₃H₁₀ClFN₂O
Storage Temperature: Ambient
MDL Number: MFCD00018540
CAS Number: 404-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR