Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Supplier: Apollo Scientific
Warning

PC300986-1G PC300986-250MG
APOSPC300986-1GEA 130 EUR
APOSPC300986-1G APOSPC300986-250MG
Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈BrF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19704051
CAS Number: 1121051-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR