4-(Dimethylamino)-3-nitrobenzotrifluoride 97%

Supplier: Apollo Scientific
Warning

PC3009M-1G
APOSPC3009M-1GEA 23.2 EUR
APOSPC3009M-1G
4-(Dimethylamino)-3-nitrobenzotrifluoride 97%
4-(Dimethylamino)-3-nitrobenzotrifluoride
Formula: C₉H₉F₃N₂O₂
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236670
CAS Number: 40700-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR