Ethyl 8-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Supplier: Apollo Scientific

PC301044-1G PC301044-250MG
APOSPC301044-1GEA 522 EUR
APOSPC301044-1G APOSPC301044-250MG
Ethyl 8-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₁H₉ClF₂N₂O₂
Melting Pt: 128…130 °C
Storage Temperature: Ambient
CAS Number: 2451256-47-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR