3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid 98%

Supplier: Apollo Scientific
Warning

PC302043-1G PC302043-5G PC302043-10G
APOSPC302043-1GEA 87 EUR
APOSPC302043-1G APOSPC302043-5G APOSPC302043-10G
3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid 98%
3-[3-Fluoro-5-(trifluoromethyl)phenyl]propionic acid
Formula: C₁₀H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD09025358
CAS Number: 916420-41-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR