5'-Fluoro-2'-methylacetophenone 98%

Supplier: Apollo Scientific

PC302118-25G PC302118-5G PC302118-1G
APOSPC302118-25GEA 359 EUR
APOSPC302118-25G APOSPC302118-5G APOSPC302118-1G
5'-Fluoro-2'-methylacetophenone 98%
5'-Fluoro-2'-methylacetophenone
Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD06660189
CAS Number: 29427-49-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR