2,6-Difluoro-3-methyl-DL-phenylalanine

Supplier: Apollo Scientific

PC302123-1G PC302123-5G
APOSPC302123-1GEA 116 EUR
APOSPC302123-1G APOSPC302123-5G
2,6-Difluoro-3-methyl-DL-phenylalanine
2,6-Difluoro-3-methyl-DL-phenylalanine
Formula: C₁₀H₁₁F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06660183
CAS Number: 1043500-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR