2-Chloro-6-fluoro-3-methyl-DL-phenylalanine

Supplier: Apollo Scientific

PC302169-1G PC302169-5G PC302169-10G
APOSPC302169-1GEA 116 EUR
APOSPC302169-1G APOSPC302169-5G APOSPC302169-10G
2-Chloro-6-fluoro-3-methyl-DL-phenylalanine
2-Chloro-6-fluoro-3-methyl-DL-phenylalanine
Formula: C₁₀H₁₁ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD06660171
CAS Number: 1259983-62-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR