4-Methoxy-3-(trifluoromethyl)phenol 98%

Supplier: Apollo Scientific
Warning

PC302349-1G PC302349-5G
APOSPC302349-1GEA 111 EUR
APOSPC302349-1G APOSPC302349-5G
4-Methoxy-3-(trifluoromethyl)phenol 98%
4-Methoxy-3-(trifluoromethoxy)phenol
Formula: C₈H₇F₃O₂
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD09832303
CAS Number: 53903-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR