2,5-Bis(trifluoromethyl)benzamide 97%

Supplier: Apollo Scientific
Warning

PC302377-5G PC302377-1G
APOSPC302377-5GEA 132 EUR
APOSPC302377-5G APOSPC302377-1G
2,5-Bis(trifluoromethyl)benzamide 97%
2,5-Bis(trifluoromethyl)benzamide
Formula: C₉H₅F₆NO
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD04973747
CAS Number: 53130-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR