(2-Fluoro-5-methylphenyl)acetic acid 98%

Supplier: Apollo Scientific

PC302388-1G PC302388-5G PC302388-250MG
APOSPC302388-1GEA 128 EUR
APOSPC302388-1G APOSPC302388-5G APOSPC302388-250MG
(2-Fluoro-5-methylphenyl)acetic acid 98%
(2-Fluoro-5-methylphenyl)acetic acid
Formula: C₉H₉FO₂
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD06660230
CAS Number: 203314-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR