4-Chloro-3,5-difluorobenzaldehyde 97%

Supplier: Apollo Scientific
Warning

PC302574-10G PC302574-5G PC302574-1G PC302574-25G
APOSPC302574-10GEA 582 EUR
APOSPC302574-10G APOSPC302574-5G APOSPC302574-1G APOSPC302574-25G
4-Chloro-3,5-difluorobenzaldehyde 97%
4-Chloro-3,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.5479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11553555
CAS Number: 1160573-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR