4-Chloro-3,5-difluorobenzoic acid 97%

Supplier: Apollo Scientific

PC302575-5G PC302575-1G PC302575-25G
APOSPC302575-5GEA 95.7 EUR
APOSPC302575-5G APOSPC302575-1G APOSPC302575-25G
4-Chloro-3,5-difluorobenzoic acid 97%
4-Chloro-3,5-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11554154
CAS Number: 1160573-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR