2-Chloro-4-(trifluoromethyl)benzaldehyde 97%

Supplier: Apollo Scientific
Warning

PC302673-1G PC302673-5G
APOSPC302673-1GEA 40.6 EUR
APOSPC302673-1G APOSPC302673-5G
2-Chloro-4-(trifluoromethyl)benzaldehyde 97%
2-Chloro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04112538
CAS Number: 82096-91-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR