4-Fluoro-2-methylbenzamide 98%

Supplier: Apollo Scientific

PC302751-1G
APOSPC302751-1GEA 72.5 EUR
APOSPC302751-1G
4-Fluoro-2-methylbenzamide 98%
4-Fluoro-2-methylbenzamide
Formula: C₈H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD04116010
CAS Number: 886501-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR